Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltriphenylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. B202525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B202525G 25 g
B2025250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B202525G Supplier TCI America Supplier No. B202525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1449-46-3
Molecular Formula C25H22BrP
Molecular Weight (g/mol) 433.33
MDL Number MFCD00031707
InChI Key WTEPWWCRWNCUNA-UHFFFAOYSA-M
Synonym benzyltriphenylphosphonium bromide, bromo benzyl triphenylphosphorane, benzyltriphenylphosphanium bromide, phenylmethyl triphenylphosphonium bromide, benzyl triphenylphosphonium bromide, benzyl triphenyl phosphonium bromide, phosphonium, triphenyl phenylmethyl-, bromide, acmc-1ch0e, ksc491k3n
PubChem CID 2734970
IUPAC Name benzyltriphenylphosphanium bromide
SMILES [Br-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 433.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyltriphenylphosphonium Bromide
EINECSNumber (3)-2608
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.