Learn More
Medchemexpress LLC Beperidium iodide | 86434-57-3 | 100.0% | 527.44 g/mol | C23H34IN3O3 | 25 MG
SDP
Catalog No. 5000215350
Encompass_Preferred
Beperidium iodide is an anticholinergic research reagent that functions as a competitive antagonist at acetylcholine receptors (reported pA2 = 7.93). Supplied as the iodide salt, it is intended for in vitro research applications requiring a characterized, high-purity muscarinic receptor antagonist.
  • Acts as a competitive antagonist of acetylcholine receptors (pA2 = 7.93).
  • Provided as the iodide salt for handling and formulation stability.
  • High purity suitable for research applications.
  • Available in small-mass pack sizes for laboratory use.
  • Molecular weight 527.44 g/mol; CAS number 86434-57-3.
Catalog No. 50-002-15350 Supplier Medchemexpress LLC Supplier No. HY10015225MG
Beperidium iodide is an anticholinergic research reagent that functions as a competitive antagonist at acetylcholine receptors (reported pA2 = 7.93). Supplied as the iodide salt, it is intended for in vitro research applications requiring a characterized, high-purity muscarinic receptor antagonist.
  • Acts as a competitive antagonist of acetylcholine receptors (pA2 = 7.93).
  • Provided as the iodide salt for handling and formulation stability.
  • High purity suitable for research applications.
  • Available in small-mass pack sizes for laboratory use.
  • Molecular weight 527.44 g/mol; CAS number 86434-57-3.