Learn More
Medchemexpress LLC Beperidium iodide | 86434-57-3 | MFCD00866899 | 100.0% | 527.44 g/mol | C23H34IN3O3 | 5 MG
SDP
Catalog No. 5000263788
Encompass_Preferred
Beperidium iodide is the iodide salt of a bioactive compound used as a competitive antagonist of acetylcholine receptors. It is supplied as a small, high-purity research reagent with a reported pA2 of 7.93, intended for use as a pharmacological probe in studies of cholinergic signaling.
  • Competitive antagonist of acetylcholine receptors with reported pA2 = 7.93.
  • High purity (99.97%) suitable for analytical and experimental use.
  • Molecular formula C23H34IN3O3 and molecular weight 527.44 g/mol.
  • Available as a 5 mg research pack for laboratory studies.
  • Provided as the iodide salt for improved stability and handling.
Catalog No. 50-002-63788 Supplier Medchemexpress LLC Supplier No. HY1001525MG
Beperidium iodide is the iodide salt of a bioactive compound used as a competitive antagonist of acetylcholine receptors. It is supplied as a small, high-purity research reagent with a reported pA2 of 7.93, intended for use as a pharmacological probe in studies of cholinergic signaling.
  • Competitive antagonist of acetylcholine receptors with reported pA2 = 7.93.
  • High purity (99.97%) suitable for analytical and experimental use.
  • Molecular formula C23H34IN3O3 and molecular weight 527.44 g/mol.
  • Available as a 5 mg research pack for laboratory studies.
  • Provided as the iodide salt for improved stability and handling.