Learn More
Medchemexpress LLC Beperidium iodide | 86434-57-3 | MFCD00866899 | 99.8% | 527.44 | C23H34IN3O3 | 50 MG
SDP
Catalog No. 5000215642
Encompass_Preferred
Beperidium iodide is the iodide salt of a muscarinic acetylcholine receptor antagonist used in research to probe muscarinic receptor pharmacology. It exhibits competitive antagonism with a reported pA2 of 7.93 and is provided as a high-purity solid suitable for in vitro assays. Documentation including a data sheet and SDS are available for laboratory handling and safety.
  • Acts as a competitive antagonist at muscarinic acetylcholine receptors (pA2 7.93).
  • High-purity solid suitable for in vitro pharmacology and receptor studies.
  • Molecular formula C23H34IN3O3 and molecular weight 527.44 g/mol.
  • Available in multiple small-scale sizes for research (mg quantities).
  • Documentation available: data sheet and safety data sheet (SDS).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-15642 Supplier Medchemexpress LLC Supplier No. HY10015250MG
Beperidium iodide is the iodide salt of a muscarinic acetylcholine receptor antagonist used in research to probe muscarinic receptor pharmacology. It exhibits competitive antagonism with a reported pA2 of 7.93 and is provided as a high-purity solid suitable for in vitro assays. Documentation including a data sheet and SDS are available for laboratory handling and safety.
  • Acts as a competitive antagonist at muscarinic acetylcholine receptors (pA2 7.93).
  • High-purity solid suitable for in vitro pharmacology and receptor studies.
  • Molecular formula C23H34IN3O3 and molecular weight 527.44 g/mol.
  • Available in multiple small-scale sizes for research (mg quantities).
  • Documentation available: data sheet and safety data sheet (SDS).
  • For research use only; not for human or clinical use.