Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bephenium hydroxynaphthoate, MilliporeSigma™ Supelco™

Catalog No. 111014096 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4096 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4096 Supplier MilliporeSigma Supelco Supplier No. B565010G

missing translation for 'validMainframeMsg'

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 3818-50-6
Physical Form Neat
Linear Formula C28H29 NO4
MDL Number MFCD00078960
Synonym Benzyldimethyl(2-phenoxyethyl)ammonium 3-hydroxynaphthoate
Type Analytical Standard
Quantity 10 g
Formula Weight 443.53
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.