Learn More
Medchemexpress LLC Bepirovirsen sodium | 2563929-84-8 | 92.7% | 7344 (free acid) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY147217A1MG
Encompass_Preferred
Bepirovirsen sodium is an antisense oligonucleotide targeting all HBV messenger RNAs. It leads to reductions in HBV-derived RNAs, HBV DNA, and viral proteins. Bepirovirsen sodium can be used for the research of chronic HBV infection.
  • Targets all HBV messenger RNAs
  • Reduces HBV-derived RNAs, HBV DNA, and viral proteins
  • Useful for chronic HBV infection research
  • Reduces HBV RNA transcripts in HBV-expressing HepG cells (in vitro)
  • Dose-dependently reduced hepatic expression of HBV RNA and cytoplasmic hepatitis B core antigen levels in HBV-transgenic mice (in vivo)
Catalog No. 50-003-62196