Learn More
Medchemexpress LLC HY-A0015 50mg Medchemexpress, Bepotastine (besilate) CAS:190786-44-8 Purity:>98%
SDP
Catalog No. 502082303
Encompass_Preferred
Medchemexpress, HY-A0015 50mg Bepotastine (besilate) CAS:190786-44-8 Bepotastine besilate is a selective and orally active second-generation histamine H1 receptor antagonist. Bepotastine besilate has the potential for allergic rhinitis, allergic conjunctivitis and urticaria/pruritus research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2303 Supplier Medchemexpress LLC Supplier No. HYA001550MG
Medchemexpress, HY-A0015 50mg Bepotastine (besilate) CAS:190786-44-8 Bepotastine besilate is a selective and orally active second-generation histamine H1 receptor antagonist. Bepotastine besilate has the potential for allergic rhinitis, allergic conjunctivitis and urticaria/pruritus research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.