Learn More
Medchemexpress LLC HY-103315 10mg Medchemexpress, Bepridil hydrochloride CAS:68099-86-5 Purity:>98%
SDP
Catalog No. 501968072
Encompass_Preferred
Medchemexpress, HY-103315 10mg Bepridil HCl CAS:68099-86-5 Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8072 Supplier Medchemexpress LLC Supplier No. HY10331510MG
Medchemexpress, HY-103315 10mg Bepridil HCl CAS:68099-86-5 Bepridil hydrochloride (CERM 1978) is a calcium channel blocker, with antianginal activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.