Learn More
Enzo Life Sciences Bepridil . hydrochloride (10mg). CAS: 74764-40-2
SDP
Supplier:  Enzo Life Sciences ALX550237M010
Encompass_Preferred
Non-selective Ca2+ channel blocker. Stimulates the binding of 1,4-dihydropyridine-based drugs to Ca2+ channels. Inhibits Na+/Ca2+ exchange. Alternative name: 1-Isobutoxy-2-pyrrolidino-3-(N-benzylanilino)propane . HCl. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9504