Learn More
Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | 99.98% | 403.00 | 1MG
SDP
Catalog No. 5000391133
Encompass_Preferred
Bepridil hydrochloride (CERM 1978) is a calcium channel blocker with antianginal activity. It is intended for research use only. In vivo, it reduces heart rate and mean arterial pressure, decreases mean coronary vascular resistance, and increases stroke volume in rats.
  • Functions as a calcium channel blocker
  • Possesses antianginal activity
  • Reduces heart rate and mean arterial pressure
  • Decreases mean coronary vascular resistance
  • Increases stroke volume
  • For research applications only
Catalog No. 50-003-91133 Supplier Medchemexpress LLC Supplier No. HY16952A1MG
Bepridil hydrochloride (CERM 1978) is a calcium channel blocker with antianginal activity. It is intended for research use only. In vivo, it reduces heart rate and mean arterial pressure, decreases mean coronary vascular resistance, and increases stroke volume in rats.
  • Functions as a calcium channel blocker
  • Possesses antianginal activity
  • Reduces heart rate and mean arterial pressure
  • Decreases mean coronary vascular resistance
  • Increases stroke volume
  • For research applications only