Learn More
Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | MFCD00065498 | 100.0% | 403.0 g/mol | C24H35ClN2O | 1 ML
SDP
Catalog No. 5000247753
Encompass_Preferred
Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker used in research to probe cardiac ion channels and antianginal mechanisms. Supplied for research and analytical applications, it is available as a solid and as a prepared solution.
  • Non-selective calcium channel blocker used in electrophysiology and pharmacology studies.
  • Hydrochloride salt form enhances solubility for assays.
  • Available as solid and as a 10 mM solution in DMSO for ready-to-use experiments.
  • High purity (~99.98%) appropriate for analytical standards.
  • Well-characterized identifiers: CAS 68099-86-5; molecular weight 403.0 g/mol.
Catalog No. 50-002-47753 Supplier Medchemexpress LLC Supplier No. HY10331510MM 1ML
Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker used in research to probe cardiac ion channels and antianginal mechanisms. Supplied for research and analytical applications, it is available as a solid and as a prepared solution.
  • Non-selective calcium channel blocker used in electrophysiology and pharmacology studies.
  • Hydrochloride salt form enhances solubility for assays.
  • Available as solid and as a 10 mM solution in DMSO for ready-to-use experiments.
  • High purity (~99.98%) appropriate for analytical standards.
  • Well-characterized identifiers: CAS 68099-86-5; molecular weight 403.0 g/mol.