Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | MFCD00065498 | 100.0% | 403.00 g/mol | C24H35ClN2O | 100 MG
SDP

Catalog No. 5000244660
Encompass_Preferred

Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker with reported antianginal activity. Supplied as a white to light yellow solid, it is used as a research reagent in cardiovascular and electrophysiology studies.

  • Cas number 68099-86-5
  • Chemical formula C24H35ClN2O
  • Molecular weight 403.00 g/mol
  • High purity suitable for research applications
  • Soluble in DMSO (100 mg/mL) and in water with ultrasonic assistance
  • Store sealed, away from moisture; in solution store at -80°C for long-term stability
  • Available in multiple pack sizes including 100 mg

Catalog No. 50-002-44660 Supplier Medchemexpress LLC Supplier No. HY103315100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker with reported antianginal activity. Supplied as a white to light yellow solid, it is used as a research reagent in cardiovascular and electrophysiology studies.

  • Cas number 68099-86-5
  • Chemical formula C24H35ClN2O
  • Molecular weight 403.00 g/mol
  • High purity suitable for research applications
  • Soluble in DMSO (100 mg/mL) and in water with ultrasonic assistance
  • Store sealed, away from moisture; in solution store at -80°C for long-term stability
  • Available in multiple pack sizes including 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.