Learn More
Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | MFCD00065498 | 100.0% | 403.00 g/mol | C24H35ClN2O | 5 MG
SDP
Catalog No. 5000244940
Encompass_Preferred
Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker with antianginal activity supplied as a research reagent for in vitro and analytical applications.
  • High purity solid (99.98%) suitable for analytical and in vitro research.
  • Non-selective calcium channel blocker, useful for cardiovascular and ion-channel studies.
  • Available in small analytical pack sizes for assay development and screening.
  • Stable when stored sealed and away from moisture; recommended -20°C for long-term storage.
  • Also available as a ready-to-use solution (10 mM in DMSO) for assay workflows.
Catalog No. 50-002-44940 Supplier Medchemexpress LLC Supplier No. HY1033155MG
Bepridil hydrochloride is the hydrochloride salt of bepridil, a non-selective calcium channel blocker with antianginal activity supplied as a research reagent for in vitro and analytical applications.
  • High purity solid (99.98%) suitable for analytical and in vitro research.
  • Non-selective calcium channel blocker, useful for cardiovascular and ion-channel studies.
  • Available in small analytical pack sizes for assay development and screening.
  • Stable when stored sealed and away from moisture; recommended -20°C for long-term storage.
  • Also available as a ready-to-use solution (10 mM in DMSO) for assay workflows.