Learn More
Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | MFCD00065498 | 99.8% | 403.0 g/mol | C24H35ClN2O | 25 MG
SDP
Catalog No. 5000244935
Encompass_Preferred
Bepridil hydrochloride is a non-selective calcium channel blocker with antianginal activity, commonly used in cardiovascular and ion channel research to study calcium-mediated cardiac effects. The compound is provided as the hydrochloride salt (CAS 68099-86-5) with molecular formula C24H35ClN2O and molecular weight 403.0 g/mol.
  • Non-selective calcium channel blocker used in cardiac and ion channel studies.
  • Useful for investigating calcium-dependent electrophysiology and myocardial contractility.
  • Hydrochloride salt form enhances solubility for assay preparation.
  • Intended for in vitro and ex vivo research applications.
  • Well-characterized chemical identifiers: CAS 68099-86-5; molecular formula C24H35ClN2O; molecular weight 403.0 g/mol.
Catalog No. 50-002-44935 Supplier Medchemexpress LLC Supplier No. HY10331525MG
Bepridil hydrochloride is a non-selective calcium channel blocker with antianginal activity, commonly used in cardiovascular and ion channel research to study calcium-mediated cardiac effects. The compound is provided as the hydrochloride salt (CAS 68099-86-5) with molecular formula C24H35ClN2O and molecular weight 403.0 g/mol.
  • Non-selective calcium channel blocker used in cardiac and ion channel studies.
  • Useful for investigating calcium-dependent electrophysiology and myocardial contractility.
  • Hydrochloride salt form enhances solubility for assay preparation.
  • Intended for in vitro and ex vivo research applications.
  • Well-characterized chemical identifiers: CAS 68099-86-5; molecular formula C24H35ClN2O; molecular weight 403.0 g/mol.