Learn More
Medchemexpress LLC Bepridil hydrochloride | 68099-86-5 | MFCD00868313 | 99.8% | 403.0 g/mol | C24H35ClN2O | 50 MG
SDP
Catalog No. 5000247315
Encompass_Preferred
Bepridil hydrochloride is the mono-hydrochloride salt of bepridil, a non-selective calcium channel blocker with antianginal activity. This product is offered as an analytical standard for research and analytical applications.
  • Calcium channel blocker, with antianginal activity.
  • Intended for research and analytical use.
  • High purity (reported ≈99.8%).
  • Hydrochloride salt form for increased solubility in polar solvents.
  • Chemical formula: C24H35ClN2O; molecular weight: 403.0 g/mol; CAS: 68099-86-5.
Catalog No. 50-002-47315 Supplier Medchemexpress LLC Supplier No. HY10331550MG
Bepridil hydrochloride is the mono-hydrochloride salt of bepridil, a non-selective calcium channel blocker with antianginal activity. This product is offered as an analytical standard for research and analytical applications.
  • Calcium channel blocker, with antianginal activity.
  • Intended for research and analytical use.
  • High purity (reported ≈99.8%).
  • Hydrochloride salt form for increased solubility in polar solvents.
  • Chemical formula: C24H35ClN2O; molecular weight: 403.0 g/mol; CAS: 68099-86-5.