Learn More
Medchemexpress LLC Bepridil hydrochloride hydrate | 74764-40-2 | 99.9% | 421.02 | 1 ML
SDP
Catalog No. 5000390595
Encompass_Preferred
Bepridil hydrochloride hydrate is a non-selective, long-acting Ca+ channel antagonist and Na+, K+ channel inhibitor. It possesses antianginal and type I antiarrhythmic effects and functions as a cardiac Na+/Ca2+ exchange (NCX1) inhibitor. It can be used for the research of cardiovascular disorders.
  • Blocks Ca2+-dependent action potentials in vascular smooth muscle.
  • Inhibits Ca currents and Na currents in cultured ventricular cells.
  • Decreases IKs under blockade of IKr.
  • Suitable for research into cardiovascular disorders.
Catalog No. 50-003-90595 Supplier Medchemexpress LLC Supplier No. HY16952A10MM/1ML
Bepridil hydrochloride hydrate is a non-selective, long-acting Ca+ channel antagonist and Na+, K+ channel inhibitor. It possesses antianginal and type I antiarrhythmic effects and functions as a cardiac Na+/Ca2+ exchange (NCX1) inhibitor. It can be used for the research of cardiovascular disorders.
  • Blocks Ca2+-dependent action potentials in vascular smooth muscle.
  • Inhibits Ca currents and Na currents in cultured ventricular cells.
  • Decreases IKs under blockade of IKr.
  • Suitable for research into cardiovascular disorders.