Learn More
Medchemexpress LLC (±)-Bepridil hydrochloride hydrate | 74764-40-2 | 99.9% | 421.02 | 25 MG
SDP
Catalog No. 5000392547
Encompass_Preferred
Bepridil hydrochloride hydrate is a non-selective, long-acting calcium channel antagonist and sodium, potassium channel inhibitor. It exhibits antianginal and type I antiarrhythmic effects, and also functions as a cardiac Na+/Ca2+ exchange (NCX1) inhibitor. This product is intended for research on cardiovascular disorders.
  • Non-selective calcium channel antagonist
  • Long-acting
  • Sodium channel inhibitor
  • Potassium channel inhibitor
  • Antianginal effects
  • Type I antiarrhythmic effects
  • Cardiac Na+/Ca2+ exchange (NCX1) inhibitor
  • Blockades of Ca2+-dependent action potentials in vascular smooth muscle
  • Blocks Ca currents and Na currents
  • Decreases IKs under blockade of IKr
  • Exhibits a half-life of averages 33±15 hours after single-dose administration
Catalog No. 50-003-92547 Supplier Medchemexpress LLC Supplier No. 25MGHY16952A
Bepridil hydrochloride hydrate is a non-selective, long-acting calcium channel antagonist and sodium, potassium channel inhibitor. It exhibits antianginal and type I antiarrhythmic effects, and also functions as a cardiac Na+/Ca2+ exchange (NCX1) inhibitor. This product is intended for research on cardiovascular disorders.
  • Non-selective calcium channel antagonist
  • Long-acting
  • Sodium channel inhibitor
  • Potassium channel inhibitor
  • Antianginal effects
  • Type I antiarrhythmic effects
  • Cardiac Na+/Ca2+ exchange (NCX1) inhibitor
  • Blockades of Ca2+-dependent action potentials in vascular smooth muscle
  • Blocks Ca currents and Na currents
  • Decreases IKs under blockade of IKr
  • Exhibits a half-life of averages 33±15 hours after single-dose administration