Learn More
Cayman Chemical BeprIdIlhydrochlorIde 10mg
SDP
Catalog No. 502983803
Encompass_Preferred
A dihydropyridine that has anti-anginal and anti-arrhythmic actions by blocking calcium channels in heart and smooth muscle; recognized as a non-selective channel blocker
Catalog No. 50-298-3803 Supplier Cayman Chemical Supplier No. 1964510 MG
A dihydropyridine that has anti-anginal and anti-arrhythmic actions by blocking calcium channels in heart and smooth muscle; recognized as a non-selective channel blocker