Learn More
Cayman Chemical Bepridil (hydrochloride), Purity: ≥98%, CAS: 68099-86-5, 50 mg
SDP
Catalog No. 502983323
Encompass_Preferred
A dihydropyridine that has anti-anginal and anti-arrhythmic actions by blocking calcium channels in heart and smooth muscle; recognized as a non-selective channel blocker
Catalog No. 50-298-3323 Supplier Cayman Chemical Supplier No. 1964550 MG
A dihydropyridine that has anti-anginal and anti-arrhythmic actions by blocking calcium channels in heart and smooth muscle; recognized as a non-selective channel blocker