Learn More
Biocare Medical, LLC Ber-EP4 Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM107AA
Encompass_Preferred
Ber-EP4 is present on the surface and the cytoplasm of all epithelial cells except the superficial layers of squamous epithelial, hepatocytes and parietal cells. It shows a broad spectrum of reactivity with human epithelial cells including simple epithelia and basal layers of stratified non-keratinized squamous epithelium and epidermis. It does not label mesothelial cells and rarely marks mesotheliomas and has been reported to distinguish between adenocarcinomas from pleural mesotheliomas. Studies also suggest it may be useful for differentiating basal cell carcinoma from other dermatological conditions.
Catalog No. 50-830-53