Learn More
Medchemexpress LLC Beraprost sodium | 88475-69-8 | 1mg
SDP
Catalog No. 5000270053
Encompass_Preferred
(Rac)-Beraprost ((Rac)-ML 1129) sodium is a racemic isomer of Beraprost sodium (HY-13569A) Beraprost sodium is an orally active prostacyclin analog that inhibits the release of Ca2 from intracellular storage sites by binding to prostacyclin membrane receptors (Prostaglandin Receptor) leading to relaxation of smooth muscle cells and vasodilation Beraprost sodium has vasodilatory antiplatelet and cytoprotective effects making it promising for research in the field of cardiovascular diseases such as thromboangiitis obliterans and atherosclerosis[1][2]
Catalog No. 50-002-70053 Supplier Medchemexpress LLC Supplier No. HYZ77211MG
(Rac)-Beraprost ((Rac)-ML 1129) sodium is a racemic isomer of Beraprost sodium (HY-13569A) Beraprost sodium is an orally active prostacyclin analog that inhibits the release of Ca2 from intracellular storage sites by binding to prostacyclin membrane receptors (Prostaglandin Receptor) leading to relaxation of smooth muscle cells and vasodilation Beraprost sodium has vasodilatory antiplatelet and cytoprotective effects making it promising for research in the field of cardiovascular diseases such as thromboangiitis obliterans and atherosclerosis[1][2]