Learn More
Selleck Chemical LLC Berbamine S9141-25MG
SDP
Catalog No. 5000024520
Encompass_Preferred
Berbamine (BA) a traditional Chinese medicines extracted from Berberis amurensis (xiaoboan) is a novel inhibitor of bcr/abl fusion gene with potent anti-leukemia activity and also an inhibitor of NF- B Berbamine (BA) induces apoptosis in human myeloma cells and inhibits the growth of cancer cells by targeting Ca /calmodulin-dependent protein kinase II (CaMKII)
Catalog No. 50-000-24520 Supplier Selleck Chemical LLC Supplier No. S914125MG
Berbamine (BA) a traditional Chinese medicines extracted from Berberis amurensis (xiaoboan) is a novel inhibitor of bcr/abl fusion gene with potent anti-leukemia activity and also an inhibitor of NF- B Berbamine (BA) induces apoptosis in human myeloma cells and inhibits the growth of cancer cells by targeting Ca /calmodulin-dependent protein kinase II (CaMKII)