Learn More
Selleck Chemical LLC Berbamine dihydrochloride S3609-1g
SDP
Supplier:  Selleck Chemical LLC S36091G
Encompass_Preferred
Berbamine (BA BBM) dihydrochloride a traditional Chinese medicines extracted from Berberis amurensis (xiaoboan) is a novel inhibitor of bcr/abl fusion gene with potent anti-leukemia activity and also an inhibitor of NF- B Berbamine (BA BBM) dihydrochloride induces apoptosis in human myeloma cells and inhibits the growth of cancer cells by targeting Ca /calmodulin-dependent protein kinase II (CaMKII)
Catalog No. 50-311-0751