Learn More
Cayman Chemical Berberine chloride
SDP
Supplier:  Cayman Chemical 100064271 G
Encompass_Preferred
An alkoloid natural product with hypocholesterolemic activity; reduces total cholesterol, LDL cholesterol, and triglycerides in humans and hamsters; enhances LDL-receptor protein and mRNA levels in hepatocytes
Catalog No. 50-288-9305