Learn More
Selleck Chemical LLC Berberine chloride hydrate
SDP
Catalog No. 5000003413
Encompass_Preferred
Berberine (Natural Yellow 18) chloride hydrate is a quaternary ammonium salt from the group of isoquinoline alkaloids Berberine activates caspase 3 and caspase 8 cleavage of poly ADP-ribose polymerase (PARP) and the release of cytochrome c Berberine chloride decreases the expression of c-IAP1 Bcl-2 and Bcl-XL Berberine chloride induces apoptosis with sustained phosphorylation of JNK and p38 MAPK as well as generation of the ROS Berberine chloride is a dual topoisomerase I and II inhibitor Berberine chloride is also a potential autophagy modulator
Catalog No. 50-000-03413 Supplier Selleck Chemical LLC Supplier No. S596725MG
Berberine (Natural Yellow 18) chloride hydrate is a quaternary ammonium salt from the group of isoquinoline alkaloids Berberine activates caspase 3 and caspase 8 cleavage of poly ADP-ribose polymerase (PARP) and the release of cytochrome c Berberine chloride decreases the expression of c-IAP1 Bcl-2 and Bcl-XL Berberine chloride induces apoptosis with sustained phosphorylation of JNK and p38 MAPK as well as generation of the ROS Berberine chloride is a dual topoisomerase I and II inhibitor Berberine chloride is also a potential autophagy modulator