Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berberine Chloride, Spectrum™ Chemical
SDP

Catalog No. 18620339 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-339 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-339 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B277725GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Natural Yellow 18

Chemical Identifiers

CAS 633-65-8

Specifications

Quantity 25 g
Molecular Weight (g/mol) 371.1
Percent Purity 97 to 102%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Berberine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.