Learn More
Selleck Chemical LLC Berberine chloride 50mg 633-65-8
SDP
Supplier:  Selleck Chemical LLC S227150MG
Encompass_Preferred
Berberine chloride is a quaternary ammonium salt from the group of isoquinoline alkaloids. Berberine activates caspase 3 and caspase 8, cleavage of poly ADP-ribose polymerase (PARP) and the release of cytochrome c. Berberine chloride decreases the expression of c-IAP1, Bcl-2 and Bcl-XL. Berberine chloride induces apoptosis with sustained phosphorylation of JNK and p38 MAPK, as well as generation of the ROS. Berberine chloride is a dual topoisomerase I and II inhibitor. Berberine chloride is also a potential autophagy modulator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-582-7