Learn More
Medchemexpress LLC Berberine hemisulfate | 316-41-6 | 384.40 | 1 G
SDP
Catalog No. 5000415889
Encompass_Preferred
Berberine hemisulfate is an alkaloid isolated from the Chinese herbal medicine Huanglian. This compound demonstrates anti-inflammatory, antibiobic, antitumor, cardiovascular protective, and neuroprotective activities, and is intended for research use only.
  • Exhibits antimicrobial activity by inhibiting cell division protein FtsZ or through DNA/RNA binding.
  • Shows anti-inflammatory activity by inhibiting TNF-α and its downstream pathways AP-1 and NF-kB.
  • Provides neuroprotective efficacy by inhibiting reactive oxygen species (ROS) production and caspase activation.
  • Attenuates metabolic diseases by regulating lipid composition and inhibiting insulin resistance.
Catalog No. 50-004-15889 Supplier Medchemexpress LLC Supplier No. HYN0716A1G
Berberine hemisulfate is an alkaloid isolated from the Chinese herbal medicine Huanglian. This compound demonstrates anti-inflammatory, antibiobic, antitumor, cardiovascular protective, and neuroprotective activities, and is intended for research use only.
  • Exhibits antimicrobial activity by inhibiting cell division protein FtsZ or through DNA/RNA binding.
  • Shows anti-inflammatory activity by inhibiting TNF-α and its downstream pathways AP-1 and NF-kB.
  • Provides neuroprotective efficacy by inhibiting reactive oxygen species (ROS) production and caspase activation.
  • Attenuates metabolic diseases by regulating lipid composition and inhibiting insulin resistance.