Learn More
Medchemexpress LLC Berberine hemisulfate | 316-41-6 | 384.40 | 100 MG
SDP
Catalog No. 5000416702
Encompass_Preferred
Berberine hemisulfate is the hemisulfate form of Berberine, an alkaloid from the Chinese herbal medicine Huanglian. It exhibits anti-inflammatory, antibiobic, antitumor, cardiovascular protective, and neuroprotective activity.
  • Exhibits antimicrobial activity by inhibiting cell division protein FtsZ or through DNA/RNA binding and damage.
  • Shows anti-inflammatory activity by inhibiting TNF-α and activation of its downstream pathway AP-1 and NF-kB.
  • Demonstrates neuroprotective efficacy by inhibiting reactive oxygen species (ROS) production and caspase activation.
  • Attenuates metabolic diseases by regulating lipid composition and inhibiting insulin resistance.
Catalog No. 50-004-16702 Supplier Medchemexpress LLC Supplier No. HYN0716A100MG
Berberine hemisulfate is the hemisulfate form of Berberine, an alkaloid from the Chinese herbal medicine Huanglian. It exhibits anti-inflammatory, antibiobic, antitumor, cardiovascular protective, and neuroprotective activity.
  • Exhibits antimicrobial activity by inhibiting cell division protein FtsZ or through DNA/RNA binding and damage.
  • Shows anti-inflammatory activity by inhibiting TNF-α and activation of its downstream pathway AP-1 and NF-kB.
  • Demonstrates neuroprotective efficacy by inhibiting reactive oxygen species (ROS) production and caspase activation.
  • Attenuates metabolic diseases by regulating lipid composition and inhibiting insulin resistance.