Learn More
TARGETMOL CHEMICALS INC BERBERINE URSODEOXYCHOLA 10MG
SDP
Catalog No. 502576450
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Berberine ursodeoxycholate (HTD1801) is an orally effective hypolipidemic agent an ionic salt of Berberine and Ursodeoxycholic acid.Berberine ursodeoxycholate has a wide range of metabolic activity and significantly reduces liver fat content. Berberine ursodeoxycholate has been used in studies of hyperlipidemia non-alcoholic steatohepatitis (NASH) and diabetes. purity: 100%
Catalog No. 50-257-6450 Supplier TARGETMOL CHEMICALS INC Supplier No. T6797410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Berberine ursodeoxycholate (HTD1801) is an orally effective hypolipidemic agent an ionic salt of Berberine and Ursodeoxycholic acid.Berberine ursodeoxycholate has a wide range of metabolic activity and significantly reduces liver fat content. Berberine ursodeoxycholate has been used in studies of hyperlipidemia non-alcoholic steatohepatitis (NASH) and diabetes. purity: 100%