Learn More
Medchemexpress LLC Berberine ursodeoxycholate | 1868138-66-2 | 99.3% | 727.93 | 1 ML
SDP
Catalog No. 5000362854
Encompass_Preferred
Berberine ursodeoxycholate (HTD1801) is an orally active, potent hypolipidemic agent. As an ionic salt of Berberine and Ursodeoxycholic acid, it significantly reduces liver fat content and exhibits broad metabolic activity. It is researched for conditions like hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes.
  • Orally active and potent hypolipidemic agent.
  • Shows significantly great reduction in liver fat content.
  • Has a broad spectrum of metabolic activity.
  • Can be used for the research of hyperlipidemia, non-alcoholic steatohepatitis (NASH) and diabetes.
  • Shows significant reductions in HbA1c (hemoglobin A1c) levels.
  • Promptly dissociates within the gastrointestinal tract, allowing differential absorption of Berberine and Ursodeoxycholic acid.
Catalog No. 50-003-62854 Supplier Medchemexpress LLC Supplier No. HY14724610MM/1ML
Berberine ursodeoxycholate (HTD1801) is an orally active, potent hypolipidemic agent. As an ionic salt of Berberine and Ursodeoxycholic acid, it significantly reduces liver fat content and exhibits broad metabolic activity. It is researched for conditions like hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes.
  • Orally active and potent hypolipidemic agent.
  • Shows significantly great reduction in liver fat content.
  • Has a broad spectrum of metabolic activity.
  • Can be used for the research of hyperlipidemia, non-alcoholic steatohepatitis (NASH) and diabetes.
  • Shows significant reductions in HbA1c (hemoglobin A1c) levels.
  • Promptly dissociates within the gastrointestinal tract, allowing differential absorption of Berberine and Ursodeoxycholic acid.