Learn More
Medchemexpress LLC Berberine ursodeoxycholate | 1868138-66-2 | 99.3% | 727.93 | 100 MG
SDP
Catalog No. 5000360791
Encompass_Preferred
Berberine ursodeoxycholate is an ionic salt of Berberine and Ursodeoxycholic acid. It is an orally active and potent hypolipidemic agent that significantly reduces liver fat content. This compound exhibits a broad spectrum of metabolic activity.
  • Orally active hypolipidemic agent
  • Significantly reduces liver fat content
  • Exhibits a broad spectrum of metabolic activity
  • Can be utilized for research into hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes
Catalog No. 50-003-60791 Supplier Medchemexpress LLC Supplier No. HY147246100MG
Berberine ursodeoxycholate is an ionic salt of Berberine and Ursodeoxycholic acid. It is an orally active and potent hypolipidemic agent that significantly reduces liver fat content. This compound exhibits a broad spectrum of metabolic activity.
  • Orally active hypolipidemic agent
  • Significantly reduces liver fat content
  • Exhibits a broad spectrum of metabolic activity
  • Can be utilized for research into hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes