Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | MFCD01658905 | 99.3% | 357.79 | C19H16ClNO4 | 25MG
SDP
Catalog No. 5000234957
Encompass_Preferred
Berberrubine chloride is an orally active metabolite of berberine provided for research and analytical use. It is a solid compound with molecular formula C19H16ClNO4, molecular weight 357.79, and high reported purity; preclinical studies describe anti-inflammatory activity. Recommended storage is sealed and protected from moisture and light, with short-term cold storage conditions specified for solvent solutions.
Catalog No. 50-002-34957 Supplier Medchemexpress LLC Supplier No. HY12585025MG
Berberrubine chloride is an orally active metabolite of berberine provided for research and analytical use. It is a solid compound with molecular formula C19H16ClNO4, molecular weight 357.79, and high reported purity; preclinical studies describe anti-inflammatory activity. Recommended storage is sealed and protected from moisture and light, with short-term cold storage conditions specified for solvent solutions.