Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | MFCD01658905 | 99.3% | 357.79 g/mol | C19H16ClNO4 | 10MM 1ML
SDP
Catalog No. 5000235555
Encompass_Preferred
Berberrubine chloride (CAS 15401-69-1) is an active metabolite of berberine with reported anti-inflammatory, anti-tumor, and antiviral properties. Supplied for research use as a dry solid or as a ready-to-use DMSO stock solution (10 mM, 1 mL). Typical supplied purity is about 99.3%, and analytical support documents are provided.
Catalog No. 50-002-35555 Supplier Medchemexpress LLC Supplier No. HY12585010MM 1ML
Berberrubine chloride (CAS 15401-69-1) is an active metabolite of berberine with reported anti-inflammatory, anti-tumor, and antiviral properties. Supplied for research use as a dry solid or as a ready-to-use DMSO stock solution (10 mM, 1 mL). Typical supplied purity is about 99.3%, and analytical support documents are provided.