Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | MFCD01658905 | 99.3% | 357.79 g/mol | C19H16ClNO4 | 50MG
SDP
Catalog No. 5000239517
Encompass_Preferred
Berberrubine chloride is the chloride salt of berberrubine, an isoquinoline alkaloid and an active metabolite of berberine. Supplied as a solid reagent for research and analytical applications, it is commonly used in pharmacological, metabolic, and inflammation model studies. Product identifiers include CAS 15401-69-1, molecular formula C19H16ClNO4, and high purity appropriate for laboratory use.
  • High purity (99.3%) suitable for analytical work.
  • Verified molecular formula C19H16ClNO4 and molecular weight 357.79 g/mol.
  • Intended for pharmacological and metabolic research applications.
  • Supplied as a solid reference material for assays and profiling.
  • Typical uses include inflammation model and metabolite studies.
Catalog No. 50-002-39517 Supplier Medchemexpress LLC Supplier No. HY12585050MG
Berberrubine chloride is the chloride salt of berberrubine, an isoquinoline alkaloid and an active metabolite of berberine. Supplied as a solid reagent for research and analytical applications, it is commonly used in pharmacological, metabolic, and inflammation model studies. Product identifiers include CAS 15401-69-1, molecular formula C19H16ClNO4, and high purity appropriate for laboratory use.
  • High purity (99.3%) suitable for analytical work.
  • Verified molecular formula C19H16ClNO4 and molecular weight 357.79 g/mol.
  • Intended for pharmacological and metabolic research applications.
  • Supplied as a solid reference material for assays and profiling.
  • Typical uses include inflammation model and metabolite studies.