Learn More
TARGETMOL CHEMICALS INC BERBERRUBINE CHLORIDE 20MG
SDP
Catalog No. 502576720
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Berberrubine chloride (9-Berberoline Chloride) has antitumor activity. 2. Berberrubine has antidiabetic activity. 3. Berberrubine dose-dependently inhibits IL-8 and MCP-1 protein levels in the media and mRNA expression of the cells stimulated with IL-1beta or TNF-alpha. purity: 97%
Catalog No. 50-257-6720 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S079520MG
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Berberrubine chloride (9-Berberoline Chloride) has antitumor activity. 2. Berberrubine has antidiabetic activity. 3. Berberrubine dose-dependently inhibits IL-8 and MCP-1 protein levels in the media and mRNA expression of the cells stimulated with IL-1beta or TNF-alpha. purity: 97%