Learn More
Medchemexpress LLC HY-N0370 1g Medchemexpress, Bergapten CAS:484-20-8 Purity:>98%
SDP
Catalog No. 501933540
Encompass_Preferred
Medchemexpress, HY-N0370 1g Bergapten CAS:484-20-8 Bergapten is a natural anti-inflammatory and anti-tumor agent isolated from bergamot essential oil, other citrus essential oils and grapefruit juice. Bergapten is inhibitory towards mouse and human CYP isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3540 Supplier Medchemexpress LLC Supplier No. HYN03701G
Medchemexpress, HY-N0370 1g Bergapten CAS:484-20-8 Bergapten is a natural anti-inflammatory and anti-tumor agent isolated from bergamot essential oil, other citrus essential oils and grapefruit juice. Bergapten is inhibitory towards mouse and human CYP isoforms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.