Learn More
Medchemexpress LLC Bergapten (5-Methoxypsoralen) | 484-20-8 | 216.19 | 500 MG
SDP
Catalog No. 5000388461
Encompass_Preferred
Bergapten is a natural anti-inflammatory and anti-tumor agent. It is an inhibitor of mouse and human Cytochrome P450 (CYP) isoforms, with varying cellular effects across different cell lines. It has been shown to decrease N-acetyltransferase activity and exert inhibitory effects on diabetes-related osteoporosis.
  • Natural anti-inflammatory and anti-tumor agent
  • Inhibitory towards mouse and human CYP isoforms
  • Decreases N-acetyltransferase (NAT) activity
  • Protects trabecular structure and decreases osteoclastogenic differentiation
  • Suppresses RANKL-RANK signaling transduction and activation of PI3K/AKT, JNK/MAPK, and NF-κB signaling pathways
Catalog No. 50-003-88461 Supplier Medchemexpress LLC Supplier No. HYN0370500MG
Bergapten is a natural anti-inflammatory and anti-tumor agent. It is an inhibitor of mouse and human Cytochrome P450 (CYP) isoforms, with varying cellular effects across different cell lines. It has been shown to decrease N-acetyltransferase activity and exert inhibitory effects on diabetes-related osteoporosis.
  • Natural anti-inflammatory and anti-tumor agent
  • Inhibitory towards mouse and human CYP isoforms
  • Decreases N-acetyltransferase (NAT) activity
  • Protects trabecular structure and decreases osteoclastogenic differentiation
  • Suppresses RANKL-RANK signaling transduction and activation of PI3K/AKT, JNK/MAPK, and NF-κB signaling pathways