Learn More
Med Vet International Miltex Vantage Bergh Cilia Tissue Forceps, 3.5in, Fine Serrated Jaws
Supplier:  Med Vet International MILV9181110
Encompass_Preferred
The Miltex Vantage V918-1110 Bergh cilia forceps are made from high quality Japanese stainless steel and measure 3.5' in length. They have a non-locking thumb handle and straight tips with fine serrations.
Catalog No. 50-255-9473