Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Berkessel-Katsuki Epoxidation Catalyst

Catalog No. 900768100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-076-8100MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-076-8100MG Supplier Sigma Aldrich Supplier No. 900768100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Developed by Albrecht Berkessel's lab, this is the first catalyst that allows catalytic enantioselective epoxidation of non-conjugated terminal double bonds with hydrogen peroxide as the oxidant. Eradicating multiple steps and kinetic resolution, this pre-formed chiral ligand-Ti2O2 catalyst complex provides a one-step route to enantiopure epoxides that are important intermediates for synthesis of bioactive compounds. Catalyst is recyclable under some conditions. For best results, pentafluorobenzoic acid P5360 or tetra-n-butylammonium hydrogensulfate 155837 is recommended as a co-catalyst.Titanium cis-1,2-Diaminocyclohexane Salalen Catalysts of Outstanding Activity and Enantioselectivity for the Asymmetric Epoxidation of Nonconjugated Terminal Olefins with Hydrogen Peroxide

Specifications

CAS 1394957-71-1
Molecular Formula C13H18N5O13P3 -+ xNa+
Linear Formula C13H18N5O13P3 -+ xNa+
Quantity 1 mg
Synonym (N6pATP) sodium salt; N6-Propargyl-adenosine-5'-triphosphate sodium salt
Molecular Weight (g/mol) 545.23 (free acid basis) g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.