Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ BCR™ Cleanroom Mopping System: Mop 1

Catalog No. 19037251 Shop All Berkshire Corporation Products
Change view
Click to view available options
Material:
Non-woven Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-037251 Non-woven Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-037251 Supplier Berkshire™ Supplier No. BCM.1.12

missing translation for 'validMainframeMsg'

Looped-end construction and fantail design minimize particle generation

  • Economical choice
  • Made of soft, nonabrasive, nonwoven polyester

Specifications

Product Type Cleanroom Mop Head
Material Non-woven Polyester
Autoclavable Autoclavable
Certifications/Compliance ISO Class 5
Color White
For Use With (Application) Easy removal of contamination from floors and for the distribution of disinfectants
Length (English) 20 in.
Width (English) 11.81 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.