Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ BCR™ Single-Knit Nylon Half-Finger Cleanroom Glove Liners

Catalog No. 19037610A Shop All Berkshire Corporation Products
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-037-610A Small
19-037-611C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-037-610A Supplier Berkshire™ Supplier No. BGL6.200SB
Add to Cart
Add to Cart
This item is not returnable. View return policy

Single-knit nylon construction is thin and comfortable, while the half-finger design permits tactility in addition to wicking moisture from the palms.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Half-finger design covers the palm and lower portion of the fingers for tactility
  • Single-knit nylon construction is thin and comfortable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Ideal for wet or temperature related processing
  • Suitable for use in ISO Class Cleanrooms

Specifications

Coating Material Uncoated
Color White
Size Small
Finger Style Half
For Use With (Application) Wet/Temperature-Related Processing, Preventing Contact Dermetitis and Perspiration
Glove Cuff Style Knit
Hem Color Red
Series BGL6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.