Learn More
Berkshire™ Labx™170 Cleanroom Wipers
Supplier:  Berkshire™ LB170 08D 24
Catalog No. 19-037269
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Made from 80% abaca and 20% wood pulp
  • Cleaner alternative to industrial tissues and wipers
  • Excellent for use as a wafer separator, wet bench liner, laboratory workmat, material packaging and blotter
  • Offer great sorptive capacity
  • Contain no surfactants and virtually no organic extractables
  • 100% biodegradeable
Product Suggestions
Product Suggestions