Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berlin Packaging™ Dangerous Goods Shipper

Catalog No. 11444100
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
70 mm or less
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-444-100 70 mm or less
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-444-100 Supplier Berlin Packaging™ Supplier No. HMS6640M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

For Use With (Equipment) Bottles of quantity 1kg (32 oz.) or less
Size 70 mm or less
Material Glass or Better
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.