Learn More
Medchemexpress LLC Beroterkib anhydrous | 2095719-92-7 | 99.9% | 584.04 g·mol⁻¹ | C29H31ClFN5O5 | 100MG
SDP
Catalog No. 5000239247
Encompass_Preferred
ASTX029 (beroterkib) is a small-molecule, dual-mechanism inhibitor of extracellular signal-regulated kinases ERK1 and ERK2 developed as a clinical candidate. It potently inhibits ERK catalytic activity and suppresses ERK phosphorylation, showing low-nanomolar biochemical and cellular potency and intended for preclinical research into MAPK-driven cancers. Supplied for research use only.
Catalog No. 50-002-39247 Supplier Medchemexpress LLC Supplier No. HY126288100MG
ASTX029 (beroterkib) is a small-molecule, dual-mechanism inhibitor of extracellular signal-regulated kinases ERK1 and ERK2 developed as a clinical candidate. It potently inhibits ERK catalytic activity and suppresses ERK phosphorylation, showing low-nanomolar biochemical and cellular potency and intended for preclinical research into MAPK-driven cancers. Supplied for research use only.