Learn More
Medchemexpress LLC Berotralstat (BCX7353) | 1809010-50-1 | MFCD33023491 | 99.7% | 562.6 g/mol | C30H26F4N6O | 5 MG
SDP
Catalog No. 5000263864
Encompass_Preferred
Berotralstat is a selective, orally active plasma kallikrein inhibitor used in research and clinical development, including prophylaxis studies for hereditary angioedema and investigational work on edema and glioblastoma models. This item is provided as a research-grade solid standard (5 mg) intended for analytical, preclinical, and formulation studies, with documented batch-level characterization and supporting analytical data.
  • Selective plasma kallikrein inhibitor for HAE and related research
  • Supplied as a 5 mg research-grade solid suitable for analytical use
  • High HPLC purity (reported ~99.7%) for reliable results
  • Documented certificate of analysis and batch data available
  • Suitable for biochemical assays, formulation studies, and reference standards
Catalog No. 50-002-63864 Supplier Medchemexpress LLC Supplier No. HY1091275MG
Berotralstat is a selective, orally active plasma kallikrein inhibitor used in research and clinical development, including prophylaxis studies for hereditary angioedema and investigational work on edema and glioblastoma models. This item is provided as a research-grade solid standard (5 mg) intended for analytical, preclinical, and formulation studies, with documented batch-level characterization and supporting analytical data.
  • Selective plasma kallikrein inhibitor for HAE and related research
  • Supplied as a 5 mg research-grade solid suitable for analytical use
  • High HPLC purity (reported ~99.7%) for reliable results
  • Documented certificate of analysis and batch data available
  • Suitable for biochemical assays, formulation studies, and reference standards