Learn More
Medchemexpress LLC Bersiporocin | 2241808-52-4 | 99.0% | C15H19Cl2N3O | 1 MG
SDP
Catalog No. 5000355244
Encompass_Preferred
Bersiporocin is a prolyl-tRNA synthetase inhibitor used in antifibrotic research. It exhibits an IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase (PRS) and is supplied as a white to off-white solid.
  • Prolyl-tRNA synthetase inhibitor
  • IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase (PRS)
  • Suitable for antifibrotic research
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture and light
  • For research use only
Catalog No. 50-003-55244 Supplier Medchemexpress LLC Supplier No. HY1455551MG
Bersiporocin is a prolyl-tRNA synthetase inhibitor used in antifibrotic research. It exhibits an IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase (PRS) and is supplied as a white to off-white solid.
  • Prolyl-tRNA synthetase inhibitor
  • IC50 of ≤100 nM for phosphoribosylpyrophosphate synthetase (PRS)
  • Suitable for antifibrotic research
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture and light
  • For research use only