Learn More
MSC Bertech Finger Cots, Latex, Powder-Free, Large, 1440/Pack
Catalog No. 5000782591
Encompass
These finger cots are designed for applications requiring protection of small parts or individual fingers where full hand protection is not necessary. Made from latex and featuring a smooth, powder-free surface, they help to maintain a clean environment and prevent contamination.
  • Ideal for handling small parts
  • Minimizes effects of perspiration, body oils, salts, and hand cream
  • Keeps moisture and dirt out of cuts or stitches
  • Smooth surface finish
  • White color
Catalog No. 50-007-82591 Supplier MSC Supplier No. 18396267
These finger cots are designed for applications requiring protection of small parts or individual fingers where full hand protection is not necessary. Made from latex and featuring a smooth, powder-free surface, they help to maintain a clean environment and prevent contamination.
  • Ideal for handling small parts
  • Minimizes effects of perspiration, body oils, salts, and hand cream
  • Keeps moisture and dirt out of cuts or stitches
  • Smooth surface finish
  • White color