Learn More
MSC Bertech Finger Cots, Powder-Free, Nitrile, Small, 4.0 mil, 720 Units
Catalog No. 5000782608
Encompass
These nitrile finger cots are designed for precision handling of small parts, providing protection without the need for full gloves. They minimize contamination from perspiration, body oils, salts, and hand creams, while also protecting cuts or stitches from moisture and dirt.
  • Powder-free design
  • Made from nitrile material
  • Small size for precision tasks
  • Thickness of 4.0 mil
  • White color
Catalog No. 50-007-82608 Supplier MSC Supplier No. 18402610
These nitrile finger cots are designed for precision handling of small parts, providing protection without the need for full gloves. They minimize contamination from perspiration, body oils, salts, and hand creams, while also protecting cuts or stitches from moisture and dirt.
  • Powder-free design
  • Made from nitrile material
  • Small size for precision tasks
  • Thickness of 4.0 mil
  • White color