Learn More
MSC Bertech Floor Tape, 2 in., Black/White, 1 roll
Catalog No. 5000688595
Encompass
This floor and aisle marking tape is designed for safety warning in industrial environments such as fields, plants, and warehouses, as well as for color-coding and general floor and aisle marking. It is 2 in. wide and 6.5 mil thick, made from PVC material with a rubber adhesive. The tape comes in a black and white striped pattern and has a smooth surface, suitable for light-duty traffic areas.
  • Designed for safety warning, color-coding, and general floor and aisle marking
  • PVC material with rubber adhesive
  • Black and white striped pattern
  • Smooth surface type
  • Suitable for light-duty traffic
  • Tensile strength of 14.00 Lb./Inch
  • Roll length of 54 ft.
Catalog No. 50-006-88595 Supplier MSC Supplier No. 18393348
This floor and aisle marking tape is designed for safety warning in industrial environments such as fields, plants, and warehouses, as well as for color-coding and general floor and aisle marking. It is 2 in. wide and 6.5 mil thick, made from PVC material with a rubber adhesive. The tape comes in a black and white striped pattern and has a smooth surface, suitable for light-duty traffic areas.
  • Designed for safety warning, color-coding, and general floor and aisle marking
  • PVC material with rubber adhesive
  • Black and white striped pattern
  • Smooth surface type
  • Suitable for light-duty traffic
  • Tensile strength of 14.00 Lb./Inch
  • Roll length of 54 ft.